Assay Details
| Gene Symbol: Npat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081152.1 | NP_001074621.1 | 1 - 2 | 134 | 135 | |
| GenBank mRNA | AK033012.1 | - | 1 - 2 | 114 | 135 | |
| AK080294.1 | - | 1 - 2 | 135 | 135 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033012.1, NM_001081152.1 transcripts.
Set Membership: |
|

Npat
Genomic Map