Assay Details
| Gene Symbol: Foxred1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291448.1 | NP_001278377.1 | 5 - 6 | 800 | 91 | |
| NM_001291449.1 | NP_001278378.1 | 5 - 6 | 805 | 91 | ||
| NM_172291.2 | NP_758495.1 | 5 - 6 | 800 | 91 | ||
| GenBank mRNA | AK054231.1 | - | 5 - 6 | 692 | 91 | |
| AK144759.1 | - | 5 - 6 | 726 | 91 | ||
| AK163722.1 | - | 5 - 6 | 659 | 91 | ||
| BC017604.1 | - | 5 - 6 | 758 | 91 | ||
| BC024806.1 | - | 5 - 6 | 670 | 5361053 | 91 | |
| BC028892.1 | - | 3 - 4 | 91 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Foxred1
Genomic Map