Assay Details
| Gene Symbol: Pate4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020264.4 | NP_064660.2 | 2 - 3 | 220 | 75 | |
| GenBank mRNA | AF134204.1 | - | 2 - 3 | 181 | 75 | |
| AK020540.1 | - | 2 - 3 | 204 | 75 | ||
| BC120764.1 | - | 2 - 3 | 220 | 40129687 | 75 | |
| BC120766.1 | - | 2 - 3 | 220 | 40129689 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194293 | AF134204.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194293 | AF134204.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453766 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580190 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 135462 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194293 | AF134204.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194293 | AF134204.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26423651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26423656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26423643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26423664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26423648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26423646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26423662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26423650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26423657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26423655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26423661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26423645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26423654 | - | - |
| Whole Mouse Genome G4122A | A_51_P194293 | AF134204.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P194293 | AF134204.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119362 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07487928 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07537743 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07487929 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07537746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26423646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07917200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07487929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08094660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07749168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07917195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26423633 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10592169 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592165 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592167 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592168 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592166 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422515_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10592164 | - | - |
| Mouse Genome 430 2.0 Array | 1422515_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422515_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56872_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56872_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900016802.mm.1 | NM_020264.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016802.mm.1 | NM_020264.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016802.mm.1 | TC0900002021.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900016801.mm.1 | NM_020264.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016801.mm.1 | NM_020264.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016801.mm.1 | TC0900002021.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422515_at | - | - |
| Mouse Expression Set 430 | 1422515_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422515_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422515_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422515_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422515_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 399822 | - | - |
| ABI Mouse Genome Survey Microarray | 426001 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4730647 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004730647 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2766350 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2766350 | - | - |
| mouse-6 v1.1 (Illumina) | scl36042.3.1_45-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4730647 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36042.3.1_45-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4730647 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2797040 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2797040 | - | - |

Pate4
Genomic Map