Assay Details
| Gene Symbol: Cdon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021339.2 | NP_067314.2 | 7 - 8 | 1576 | 67 | |
| GenBank mRNA | AF090866.1 | - | 7 - 8 | 1576 | 67 | |
| AK040711.1 | - | 7 - 8 | 1399 | 67 | ||
| AK157750.1 | - | 9 - 10 | 1759 | 67 | ||
| BC109176.2 | - | 7 - 8 | 1575 | 40048829 | 67 | |
| CB526103.1 | - | 2 - 3 | 434 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdon
Genomic Map