Assay Details
| Gene Symbol: Usp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK031141.1 | - | 8 - 9 | 869 | 107 | |
| AK158516.1 | - | 6 - 7 | 684 | 107 | ||
| AK161676.1 | - | 7 - 8 | 812 | 107 | ||
| BC017156.1 | - | 8 - 9 | 990 | 4911201 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp3
Genomic Map