Assay Details
| Gene Symbol: Dapk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB018002.1 | - | 2 - 3 | 208 | 76 | |
| AK158082.1 | - | 2 - 3 | 210 | 76 | ||
| BC022165.1 | - | 2 - 3 | 187 | 4016911 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dapk2
Genomic Map