Assay Details
| Gene Symbol: Aldh1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009022.4 | NP_033048.2 | 1 - 2 | 168 | 65 | |
| GenBank mRNA | AK078553.1 | - | 1 - 2 | 109 | 65 | |
| BC075704.1 | - | 1 - 2 | 135 | 30471325 | 65 | |
| CJ166631.1 | - | 1 - 2 | 169 | 65 | ||
| X99273.1 | - | 1 - 2 | 113 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aldh1a2
Genomic Map