Assay Details
| Gene Symbol: Eif3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016876.3 | NP_058572.2 | 2 - 3 | 91 | 86 | |
| GenBank mRNA | AF108214.1 | - | 2 - 3 | 105 | 86 | |
| AK038249.1 | - | 2 - 3 | 101 | 86 | ||
| AK168362.1 | - | 2 - 3 | 94 | 86 | ||
| BC008511.1 | - | 2 - 3 | 91 | 3592537 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif3g
Genomic Map