Assay Details
| Gene Symbol: Eif3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016876.3 | NP_058572.2 | 3 - 4 | 175 | 145 | |
| GenBank mRNA | AF108214.1 | - | 3 - 4 | 189 | 145 | |
| AK038249.1 | - | 3 - 4 | 185 | 145 | ||
| AK168362.1 | - | 3 - 4 | 178 | 145 | ||
| BC008511.1 | - | 3 - 4 | 175 | 3592537 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif3g
Genomic Map