Assay Details
| Gene Symbol: Nfkbib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK010218.1 | - | 3 - 4 | 678 | 83 | |
| AK038631.1 | - | 3 - 4 | 726 | 83 | ||
| AK156731.1 | - | 3 - 4 | 652 | 83 | ||
| AY176058.1 | - | 3 - 4 | 685 | 83 | ||
| AY176059.1 | - | 3 - 4 | 716 | 83 | ||
| BC021938.1 | - | 3 - 4 | 683 | 5136809 | 83 | |
| U19799.1 | - | 3 - 4 | 697 | 83 | ||
More Information
Set Membership: |
|

Nfkbib
Genomic Map