Assay Details
| Gene Symbol: Ech1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016772.1 | NP_058052.1 | 9 - 10 | 905 | 60 | |
| GenBank mRNA | AF030343.1 | - | 9 - 10 | 905 | 60 | |
| AK088495.1 | - | 9 - 10 | 893 | 60 | ||
| BC068112.1 | - | 9 - 10 | 905 | 5710971 | 60 | |
| BC087924.1 | - | 9 - 10 | 966 | 6706361 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25023777 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25023776 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25023778 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P421846 | NM_016772.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P421846 | AF030343.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P421846 | AK088495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P421846 | BC068112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P421846 | BC087924.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P421846 | NM_016772.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P421846 | AF030343.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P421846 | AK088495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P421846 | BC068112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P421846 | BC087924.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614048 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92828 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 954663 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9967 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P421846 | NM_016772.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P421846 | AF030343.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P421846 | AK088495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P421846 | BC068112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P421846 | BC087924.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P421846 | NM_016772.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P421846 | AF030343.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P421846 | AK088495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P421846 | BC068112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P421846 | BC087924.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421846 | NM_016772.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421846 | AF030343.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421846 | AK088495.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421846 | BC068112.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421846 | BC087924.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421846 | NM_016772.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421846 | AF030343.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421846 | AK088495.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421846 | BC068112.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421846 | BC087924.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25023771 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06150496 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06150505 | - | - |
| Whole Mouse Genome G4122A | A_51_P421846 | NM_016772.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421846 | AF030343.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421846 | AK088495.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421846 | BC068112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421846 | BC087924.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421846 | NM_016772.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421846 | AF030343.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421846 | AK088495.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421846 | BC068112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421846 | BC087924.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118769 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110839 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05967882 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05967879 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06187074 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05967883 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25023778 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25023776 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25023777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25023767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06184098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06551157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06152928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05967881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06127361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06155010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05967882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06255079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25023800 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10551623 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551617 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551619 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551620 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551625 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551618 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551615 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551622 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551616 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551624 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551621 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448491_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10551614 | - | - |
| Mouse Genome 430 2.0 Array | 1448491_at | - | - |
| Mouse Genome 430 2.0 Array | 1448491_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 51798_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 51798_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700004286.mm.1 | NM_016772.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700004286.mm.1 | NM_016772.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700004286.mm.1 | TC0700000514.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448491_PM_at | - | - |
| Mouse Expression Set 430 | 1448491_at | - | - |
| Murine Genome U74v2 Set | 93754_PM_at | - | - |
| Murine Genome U74v2 Set | 93754_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448491_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448491_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448491_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448491_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 874207 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4120746 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004120746 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1254786 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254786 | - | - |
| mouse-6 v1.1 (Illumina) | scl32856.10.1_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4120746 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32856.10.1_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4120746 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1254786 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254786 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32856.10.1_9-S | - | - |

Ech1
Genomic Map