Assay Details
| Gene Symbol: Pcbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025273.4 | NP_079549.1 | 2 - 3 | 219 | 88 | |
| GenBank mRNA | AK007401.1 | - | 2 - 3 | 191 | 88 | |
| BC024354.1 | - | 2 - 3 | 152 | 5052788 | 88 | |
| M83741.1 | - | 1 - 2 | 129 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcbd1
Genomic Map