Assay Details
| Gene Symbol: Ift46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023831.3 | NP_076320.2 | 6 - 7 | 646 | 106 | |
| GenBank mRNA | AJ249981.1 | - | 6 - 7 | 690 | 106 | |
| AK005360.1 | - | 6 - 7 | 654 | 106 | ||
| AK032880.1 | - | 6 - 7 | 647 | 106 | ||
| BC010326.1 | - | 5 - 6 | 445 | 3599905 | 106 | |
| BC080764.1 | - | 6 - 7 | 588 | 6488976 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ift46
Genomic Map