Assay Details
| Gene Symbol: Ift46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023831.3 | NP_076320.2 | 8 - 9 | 900 | 85 | |
| GenBank mRNA | AJ249981.1 | - | 8 - 9 | 944 | 85 | |
| AK005360.1 | - | 8 - 9 | 908 | 85 | ||
| AK032880.1 | - | 8 - 9 | 901 | 85 | ||
| BC010326.1 | - | 7 - 8 | 699 | 3599905 | 85 | |
| BC080764.1 | - | 8 - 9 | 842 | 6488976 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ift46
Genomic Map