Assay Details
| Gene Symbol: Hmgcll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173731.2 | NP_776092.1 | 6 - 7 | 698 | 71 | |
| GenBank mRNA | AK141622.1 | - | 6 - 7 | 661 | 71 | |
| BC037381.1 | - | 6 - 7 | 698 | 5368383 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hmgcll1
Genomic Map