Assay Details
| Gene Symbol: Aagab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025857.2 | NP_080133.1 | 2 - 3 | 309 | 62 | |
| GenBank mRNA | AK017714.1 | - | 2 - 3 | 278 | 62 | |
| AK159897.1 | - | 2 - 3 | 310 | 62 | ||
| AK192186.1 | - | 1 - 2 | 101 | 62 | ||
| AK214885.1 | - | 1 - 2 | 101 | 62 | ||
| AK215257.1 | - | 1 - 2 | 101 | 62 | ||
| BC027311.1 | - | 2 - 3 | 305 | 3964905 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aagab
Genomic Map