Assay Details
| Gene Symbol: Aagab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025857.2 | NP_080133.1 | 5 - 6 | 583 | 64 | |
| GenBank mRNA | AK017714.1 | - | 5 - 6 | 552 | 64 | |
| AK159897.1 | - | 5 - 6 | 584 | 64 | ||
| BC027311.1 | - | 5 - 6 | 579 | 3964905 | 64 | |
More Information
Set Membership: |
|

Aagab
Genomic Map