Assay Details
| Gene Symbol: Psmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011970.4 | NP_036100.3 | 2 - 3 | 318 | 90 | |
| GenBank mRNA | AF060090.1 | - | 2 - 3 | 206 | 90 | |
| AK078487.1 | - | 2 - 3 | 297 | 90 | ||
| BB853639.1 | - | 2 - 3 | 331 | 90 | ||
| BC008265.1 | - | 2 - 3 | 245 | 2811010 | 90 | |
| CB321399.1 | - | 2 - 3 | 314 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmb2
Genomic Map