Assay Details
| Gene Symbol: Psmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011970.4 | NP_036100.3 | 4 - 5 | 561 | 60 | |
| GenBank mRNA | AF060090.1 | - | 4 - 5 | 449 | 60 | |
| AK078487.1 | - | 4 - 5 | 540 | 60 | ||
| BC008265.1 | - | 4 - 5 | 488 | 2811010 | 60 | |
| BQ043213.1 | - | 1 - 2 | 44 | 60 | ||
| CB321399.1 | - | 4 - 5 | 557 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmb2
Genomic Map