Assay Details
| Gene Symbol: Dlgap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK158868.1 | - | 2 - 3 | 575 | 117 | |
| AW123730.1 | - | 2 - 3 | 251 | 117 | ||
| BC057615.1 | - | 2 - 3 | 184 | 6401651 | 117 | |
| BC098311.1 | - | 778 | 117 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK158868.1, BC057615.1 transcripts.
Set Membership: |
|

Dlgap3
Genomic Map