Assay Details
| Gene Symbol: Dlgap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK158868.1 | - | 4 - 5 | 1928 | 91 | |
| BC057615.1 | - | 4 - 5 | 1537 | 6401651 | 91 | |
| BC058433.1 | - | 2 - 3 | 735 | 91 | ||
| BC098311.1 | - | 2070 | 91 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dlgap3
Genomic Map