Assay Details
| Gene Symbol: Dlgap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK158868.1 | - | 8 - 9 | 2614 | 55 | |
| AY243848.2 | - | 6 - 7 | 1994 | 55 | ||
| BC057615.1 | - | 8 - 9 | 2223 | 6401651 | 55 | |
| BC058433.1 | - | 6 - 7 | 1421 | 55 | ||
| BC098311.1 | - | 2756 | 55 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dlgap3
Genomic Map