Assay Details
| Gene Symbol: Matn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010769.2 | NP_034899.2 | 1 - 2 | 142 | 146 | |
| GenBank mRNA | BC047140.1 | - | 1 - 2 | 109 | 6315509 | 146 |
More Information
Updated Mapping Information
Set Membership: |
|

Matn1
Genomic Map