Assay Details
| Gene Symbol: Aebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF053943.1 | - | 11 - 12 | 1578 | 61 | |
| AK004712.1 | - | 7 - 8 | 587 | 61 | ||
| AK128980.1 | - | 11 - 12 | 1654 | 61 | ||
| AK158276.1 | - | 11 - 12 | 1605 | 61 | ||
| AK159330.1 | - | 11 - 12 | 1585 | 61 | ||
| AK159342.1 | - | 11 - 12 | 1604 | 61 | ||
| AK159377.1 | - | 11 - 12 | 1575 | 61 | ||
| AK159409.1 | - | 11 - 12 | 1605 | 61 | ||
| AK159774.1 | - | 11 - 12 | 1576 | 61 | ||
| AK159957.1 | - | 11 - 12 | 1576 | 61 | ||
| AK195518.1 | - | 1 - 2 | 158 | 61 | ||
| BC082577.1 | - | 11 - 12 | 1590 | 30648530 | 61 | |
| X80478.1 | - | 2 - 3 | 308 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aebp1
Genomic Map