Assay Details
| Gene Symbol: Aebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF053943.1 | - | 6 - 7 | 1111 | 102 | |
| AK004712.1 | - | 2 - 3 | 117 | 102 | ||
| AK128980.1 | - | 6 - 7 | 1187 | 102 | ||
| AK158276.1 | - | 6 - 7 | 1135 | 102 | ||
| AK159330.1 | - | 6 - 7 | 1115 | 102 | ||
| AK159342.1 | - | 6 - 7 | 1134 | 102 | ||
| AK159377.1 | - | 6 - 7 | 1105 | 102 | ||
| AK159409.1 | - | 6 - 7 | 1135 | 102 | ||
| AK159774.1 | - | 6 - 7 | 1106 | 102 | ||
| AK159957.1 | - | 6 - 7 | 1106 | 102 | ||
| BC082577.1 | - | 6 - 7 | 1123 | 30648530 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aebp1
Genomic Map