Assay Details
| Gene Symbol: Aebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF053943.1 | - | 9 - 10 | 1327 | 128 | |
| AK004712.1 | - | 5 - 6 | 336 | 131 | ||
| AK128980.1 | - | 9 - 10 | 1403 | 128 | ||
| AK158276.1 | - | 9 - 10 | 1354 | 131 | ||
| AK159330.1 | - | 9 - 10 | 1334 | 131 | ||
| AK159342.1 | - | 9 - 10 | 1353 | 131 | ||
| AK159377.1 | - | 9 - 10 | 1324 | 131 | ||
| AK159409.1 | - | 9 - 10 | 1354 | 131 | ||
| AK159774.1 | - | 9 - 10 | 1325 | 131 | ||
| AK159957.1 | - | 9 - 10 | 1325 | 131 | ||
| BC082577.1 | - | 9 - 10 | 1339 | 30648530 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aebp1
Genomic Map