Assay Details
| Gene Symbol: Ckb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021273.4 | NP_067248.1 | 6 - 7 | 906 | 92 | |
| GenBank mRNA | AK002467.1 | - | 6 - 7 | 869 | 92 | |
| AK014299.1 | - | 6 - 7 | 849 | 92 | ||
| AK148885.1 | - | 6 - 7 | 866 | 92 | ||
| AK152388.1 | - | 6 - 7 | 866 | 92 | ||
| AK153484.1 | - | 6 - 7 | 866 | 92 | ||
| AK161990.1 | - | 6 - 7 | 866 | 92 | ||
| AK166980.1 | - | 6 - 7 | 869 | 92 | ||
| AK167034.1 | - | 6 - 7 | 869 | 92 | ||
| BC015271.1 | - | 6 - 7 | 836 | 4225384 | 92 | |
| BC106109.1 | - | 6 - 7 | 833 | 6395794 | 92 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ckb
Genomic Map