Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100022638.mm.1 |
NM_001033865.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022638.mm.1 |
NM_001033865.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022638.mm.1 |
NM_024277.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022638.mm.1 |
NM_024277.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022638.mm.1 |
TC1100002421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100022642.mm.1 |
NM_001033865.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022642.mm.1 |
NM_001033865.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022642.mm.1 |
TC1100002421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100022635.mm.1 |
NM_001033865.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022635.mm.1 |
NM_001033865.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022635.mm.1 |
NM_024277.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022635.mm.1 |
NM_024277.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022635.mm.1 |
TC1100002421.mm.1 |
MTA TC |