Assay Details
| Gene Symbol: Grb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177629.1 | NP_001171100.1 | 11 - 12 | 1285 | 86 | |
| NM_010345.4 | NP_034475.2 | 12 - 13 | 1610 | 86 | ||
| GenBank mRNA | AF022072.1 | - | 12 - 13 | 1584 | 86 | |
| AK030727.1 | - | 12 - 13 | 1545 | 86 | ||
| AK032927.1 | - | 12 - 13 | 1591 | 86 | ||
| AK163841.1 | - | 12 - 13 | 1591 | 86 | ||
| AK169000.1 | - | 6 - 7 | 574 | 86 | ||
| BC016111.1 | - | 11 - 12 | 1285 | 4481345 | 86 | |
| BC053842.1 | - | 12 - 13 | 1577 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grb10
Genomic Map