Assay Details
| Gene Symbol: Grb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177629.1 | NP_001171100.1 | 6 - 7 | 800 | 105 | |
| NM_010345.4 | NP_034475.2 | 7 - 8 | 1125 | 105 | ||
| GenBank mRNA | AF022072.1 | - | 7 - 8 | 1099 | 105 | |
| AK030727.1 | - | 7 - 8 | 1060 | 105 | ||
| AK032927.1 | - | 7 - 8 | 1106 | 105 | ||
| AK163841.1 | - | 7 - 8 | 1106 | 105 | ||
| AK169000.1 | - | 1 - 2 | 89 | 105 | ||
| BC016111.1 | - | 6 - 7 | 800 | 4481345 | 105 | |
| BC053842.1 | - | 7 - 8 | 1092 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grb10
Genomic Map