Assay Details
| Gene Symbol: Tpd52l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009413.1 | NP_033439.1 | 3 - 4 | 474 | 56 | |
| GenBank mRNA | AF004428.1 | - | 3 - 4 | 474 | 56 | |
| AK131855.1 | - | 3 - 4 | 371 | 56 | ||
| BC141068.1 | - | 3 - 4 | 361 | 9055680 | 56 | |
| BC145321.1 | - | 3 - 4 | 361 | 9053876 | 56 | |
| BC145322.1 | - | 3 - 4 | 361 | 56 | ||
| BC171924.1 | - | 3 - 4 | 361 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tpd52l1
Genomic Map