Assay Details
| Gene Symbol: Cnksr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172546.2 | NP_766134.1 | 5 - 6 | 1101 | 83 | |
| GenBank mRNA | AK033015.1 | - | 5 - 6 | 1102 | 83 | |
| AK149951.1 | - | 4 - 5 | 400 | 83 | ||
| AY151137.1 | - | 5 - 6 | 1063 | 83 | ||
| BC024086.1 | - | 4 - 5 | 449 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnksr3
Genomic Map