Assay Details
| Gene Symbol: Cnksr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172546.2 | NP_766134.1 | 6 - 7 | 1222 | 75 | |
| GenBank mRNA | AK033015.1 | - | 6 - 7 | 1223 | 75 | |
| AK149951.1 | - | 5 - 6 | 521 | 75 | ||
| AY151137.1 | - | 6 - 7 | 1184 | 75 | ||
| BC024086.1 | - | 5 - 6 | 570 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnksr3
Genomic Map