Assay Details
| Gene Symbol: Dcbld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025705.3 | NP_079981.2 | 5 - 6 | 685 | 85 | |
| GenBank mRNA | AK014521.1 | - | 5 - 6 | 497 | 85 | |
| AK016485.1 | - | 5 - 6 | 685 | 85 | ||
More Information
Set Membership: |
|

Dcbld1
Genomic Map