Assay Details
| Gene Symbol: Dcbld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025705.3 | NP_079981.2 | 9 - 10 | 1006 | 129 | |
| GenBank mRNA | AK014521.1 | - | 9 - 10 | 818 | 129 | |
| BC026771.1 | - | 8 - 9 | 794 | 4014778 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dcbld1
Genomic Map