Assay Details
| Gene Symbol: Kdm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133872.2 | NP_598633.2 | 17 - 18 | 2374 | 78 | |
| GenBank mRNA | AK129170.1 | - | 17 - 18 | 2314 | 78 | |
| BC019417.1 | - | 3 - 4 | 320 | 78 | ||
| BC059885.1 | - | 17 - 18 | 2361 | 6815975 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdm1a
Genomic Map