Assay Details
| Gene Symbol: Camk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001174053.1 | NP_001167524.1 | 1 - 2 | 272 | 59 | |
| NM_001174054.1 | NP_001167525.1 | 1 - 2 | 272 | 59 | ||
| NM_007595.5 | NP_031621.3 | 1 - 2 | 272 | 59 | ||
| GenBank mRNA | AK048845.1 | - | 1 - 2 | 183 | 59 | |
| AK163698.1 | - | 1 - 2 | 89 | 59 | ||
| BC080273.1 | - | 1 - 2 | 74 | 6851995 | 59 | |
| CJ119973.1 | - | 1 - 2 | 269 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Camk2b
Genomic Map