Assay Details
| Gene Symbol: Camk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001174053.1 | NP_001167524.1 | 6 - 7 | 630 | 82 | |
| NM_001174054.1 | NP_001167525.1 | 6 - 7 | 630 | 82 | ||
| NM_007595.5 | NP_031621.3 | 6 - 7 | 630 | 82 | ||
| GenBank mRNA | AK046454.1 | - | 6 - 7 | 631 | 82 | |
| AK158794.1 | - | 3 - 4 | 391 | 82 | ||
| AK163698.1 | - | 6 - 7 | 447 | 82 | ||
| BC024803.1 | - | 6 - 7 | 446 | 82 | ||
| BC080273.1 | - | 6 - 7 | 432 | 6851995 | 82 | |
| X63615.1 | - | 6 - 7 | 445 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK158794.1 transcripts.
Set Membership: |
|

Camk2b
Genomic Map