Assay Details
| Gene Symbol: Abcg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011920.3 | NP_036050.1 | 7 - 8 | 1290 | 60 | |
| GenBank mRNA | AF140218.1 | - | 6 - 7 | 853 | 60 | |
| BC053730.1 | - | 7 - 8 | 1302 | 30064535 | 60 | |
| KF662864.1 | - | 7 - 8 | 887 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcg2
Genomic Map