Assay Details
| Gene Symbol: Morc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159288.1 | NP_001152760.1 | 20 - 21 | 2912 | 71 | |
| NM_198162.1 | NP_937805.1 | 19 - 20 | 2239 | 71 | ||
| GenBank mRNA | AK173042.1 | - | 20 - 21 | 2912 | 71 | |
| BC059243.1 | - | 19 - 20 | 2239 | 6417551 | 71 | |
| BC063082.1 | - | 8 - 9 | 1179 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Morc2a
Genomic Map