Assay Details
| Gene Symbol: Morc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159288.1 | NP_001152760.1 | 24 - 25 | 3466 | 106 | |
| NM_198162.1 | NP_937805.1 | 23 - 24 | 2796 | 106 | ||
| GenBank mRNA | AK018377.1 | - | 4 - 5 | 540 | 106 | |
| AK173042.1 | - | 24 - 25 | 3466 | 106 | ||
| BC059243.1 | - | 23 - 24 | 2796 | 6417551 | 106 | |
| BC063082.1 | - | 12 - 13 | 1733 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Morc2a
Genomic Map