Assay Details
| Gene Symbol: Smyd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160127.1 | NP_001153599.1 | 3 - 4 | 606 | 83 | |
| NM_009762.2 | NP_033892.2 | 3 - 4 | 606 | 83 | ||
| GenBank mRNA | AK076266.1 | - | 3 - 4 | 571 | 83 | |
| AK142153.1 | - | 3 - 4 | 574 | 83 | ||
| AK142252.1 | - | 3 - 4 | 584 | 83 | ||
| BC076601.1 | - | 3 - 4 | 584 | 30609878 | 83 | |
| U76371.1 | - | 83 | ||||
| U76373.2 | - | 3 - 4 | 578 | 83 | ||
| U76374.2 | - | 3 - 4 | 578 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smyd1
Genomic Map