Assay Details
| Gene Symbol: Lrig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008377.2 | NP_032403.2 | 10 - 11 | 1695 | 61 | |
| GenBank mRNA | BC141300.1 | - | 10 - 11 | 1487 | 9055912 | 61 |
| BC145623.1 | - | 10 - 11 | 1487 | 9054191 | 61 | |
| D78572.1 | - | 10 - 11 | 1688 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrig1
Genomic Map