Assay Details
| Gene Symbol: Tmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081111.2 | NP_001074580.1 | 13 - 14 | 2858 | 134 | |
| GenBank mRNA | BC150931.1 | - | 13 - 14 | 2869 | 9087842 | 134 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmf1
Genomic Map