| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600008395.mm.1 |
NM_027868.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600008395.mm.1 |
NM_027868.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600008395.mm.1 |
TC0600001078.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600008400.mm.1 |
NM_001037493.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600008400.mm.1 |
NM_027868.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600008400.mm.1 |
NM_027868.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600008400.mm.1 |
TC0600001078.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600008397.mm.1 |
NM_027868.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600008397.mm.1 |
NM_027868.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600008397.mm.1 |
TC0600001078.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600008396.mm.1 |
NM_027868.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600008396.mm.1 |
NM_027868.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600008396.mm.1 |
TC0600001078.mm.1 |
MTA TC |