Assay Details
| Gene Symbol: Slc41a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037493.2 | NP_001032570.1 | 3 - 4 | 579 | 64 | |
| NM_027868.2 | NP_082144.2 | 4 - 5 | 611 | 64 | ||
| GenBank mRNA | AK003140.1 | - | 3 - 4 | 579 | 64 | |
| BC011108.1 | - | 4 - 5 | 527 | 4011836 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc41a3
Genomic Map