Assay Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026585.3 | NP_080861.2 | 11 - 12 | 1070 | 94 | |
| GenBank mRNA | AK038318.1 | - | 11 - 12 | 1048 | 94 | |
| AK122320.1 | - | 10 - 11 | 1076 | 94 | ||
| AK145716.1 | - | 11 - 12 | 1071 | 94 | ||
| AK161181.1 | - | 11 - 12 | 1019 | 94 | ||
| AK163765.1 | - | 10 - 11 | 1058 | 94 | ||
| AK168388.1 | - | 11 - 12 | 1057 | 94 | ||
| AK198696.1 | - | 2 - 3 | 75 | 94 | ||
| BC032177.1 | - | 9 - 10 | 1297 | 94 | ||
| BC040393.1 | - | 11 - 12 | 994 | 94 | ||
| BC049979.1 | - | 11 - 12 | 1046 | 94 | ||
| BC056942.1 | - | 11 - 12 | 997 | 6827177 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam21
Genomic Map