| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600010284.mm.1 |
NM_026585.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010284.mm.1 |
NM_026585.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010284.mm.1 |
TC0600001293.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600010285.mm.1 |
NM_026585.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010285.mm.1 |
NM_026585.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010285.mm.1 |
TC0600001293.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600010283.mm.1 |
NM_026585.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010283.mm.1 |
NM_026585.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010283.mm.1 |
TC0600001293.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600010282.mm.1 |
NM_026585.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010282.mm.1 |
NM_026585.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010282.mm.1 |
TC0600001293.mm.1 |
MTA TC |