Assay Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026585.3 | NP_080861.2 | 7 - 8 | 776 | 86 | |
| GenBank mRNA | AK011156.1 | - | 7 - 8 | 722 | 86 | |
| AK038318.1 | - | 7 - 8 | 754 | 86 | ||
| AK122320.1 | - | 6 - 7 | 782 | 86 | ||
| AK145716.1 | - | 7 - 8 | 777 | 86 | ||
| AK161181.1 | - | 7 - 8 | 725 | 86 | ||
| AK163765.1 | - | 6 - 7 | 764 | 86 | ||
| AK168388.1 | - | 7 - 8 | 763 | 86 | ||
| BC032177.1 | - | 5 - 6 | 1003 | 86 | ||
| BC040393.1 | - | 7 - 8 | 700 | 86 | ||
| BC049979.1 | - | 7 - 8 | 752 | 86 | ||
| BC056942.1 | - | 7 - 8 | 703 | 6827177 | 86 | |
More Information
Set Membership: |
|

Fam21
Genomic Map