Assay Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026585.3 | NP_080861.2 | 8 - 9 | 821 | 92 | |
| GenBank mRNA | AK011156.1 | - | 8 - 9 | 767 | 92 | |
| AK038318.1 | - | 8 - 9 | 799 | 92 | ||
| AK122320.1 | - | 7 - 8 | 827 | 92 | ||
| AK145716.1 | - | 8 - 9 | 822 | 92 | ||
| AK161181.1 | - | 8 - 9 | 770 | 92 | ||
| AK163765.1 | - | 7 - 8 | 809 | 92 | ||
| AK168388.1 | - | 8 - 9 | 808 | 92 | ||
| BC032177.1 | - | 6 - 7 | 1048 | 92 | ||
| BC040393.1 | - | 8 - 9 | 745 | 92 | ||
| BC049979.1 | - | 8 - 9 | 797 | 92 | ||
| BC056942.1 | - | 8 - 9 | 748 | 6827177 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam21
Genomic Map